Assay Details
| Gene Symbol: LOC103692976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765858.1 | XP_008764080.1 | 856 | 85 | ||
| Gene Symbol: Ccnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108110.1 | NP_001101580.1 | 5 - 6 | 494 | 85 | |
| XM_008765745.1 | XP_008763967.1 | 5 - 6 | 654 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC103692976
Genomic Map