Assay Details
| Gene Symbol: Racgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108112.1 | NP_001101582.1 | 2 - 3 | 268 | 73 | |
| XM_006257400.2 | XP_006257462.1 | 2 - 3 | 215 | 73 | ||
| GenBank mRNA | BC166473.1 | - | 2 - 3 | 168 | 9090730 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Racgap1
Genomic Map