Assay Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001069014.4 | XP_001069014.1 | 4 - 5 | 318 | 147 | |
| XM_006255409.2 | XP_006255471.1 | 4 - 5 | 293 | 147 | ||
More Information
Set Membership: |
|

Gypa
Genomic Map