Assay Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001069014.4 | XP_001069014.1 | 5 - 6 | 396 | 90 | |
| XM_006255409.2 | XP_006255471.1 | 5 - 6 | 371 | 90 | ||
More Information
Set Membership: |
|

Gypa
Genomic Map