Assay Details
| Gene Symbol: Hibadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022243.1 | NP_071579.1 | 6 - 7 | 722 | 90 | |
| GenBank mRNA | BC127442.1 | - | 6 - 7 | 708 | 7308860 | 90 |
| J04628.1 | - | 6 - 7 | 723 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hibadh
Genomic Map