Assay Details
| Gene Symbol: Ildr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127536.3 | NP_001121008.2 | 2 - 3 | 310 | 61 | |
| XM_006248394.2 | XP_006248456.1 | 2 - 3 | 379 | 61 | ||
| GenBank mRNA | BC166709.1 | - | 2 - 3 | 310 | 7929589 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Ildr1
Genomic Map