Assay Details
| Gene Symbol: Rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127597.2 | NP_001121069.2 | 2 - 3 | 403 | 66 | |
| GenBank mRNA | BC166869.1 | - | 2 - 3 | 403 | 9030307 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf168
Genomic Map