Assay Details
| Gene Symbol: Tmem45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191074.1 | NP_001178003.1 | 2 - 3 | 537 | 74 | |
| Gene Symbol: Tmem45al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751037.3 | XP_003751085.1 | 2 - 3 | 629 | 74 | |
| XM_006248218.2 | XP_006248280.1 | 2 - 3 | 632 | 74 | ||
| XM_008768662.1 | XP_008766884.1 | 2 - 3 | 633 | 74 | ||
| XM_008768663.1 | XP_008766885.1 | 2 - 3 | 629 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem45a
Genomic Map