Assay Details
| Gene Symbol: Tmem45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191074.1 | NP_001178003.1 | 5 - 6 | 1068 | 131 | |
| Gene Symbol: Tmem45al | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751037.3 | XP_003751085.1 | 5 - 6 | 1160 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem45a
Genomic Map