Assay Details
| Gene Symbol: Rhox8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025776.1 | NP_001020947.1 | 3 - 4 | 485 | 69 | |
| GenBank mRNA | DQ058655.1 | - | 3 - 4 | 485 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhox8
Genomic Map