Assay Details
| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031670.2 | NP_113858.1 | 1 - 2 | 123 | 143 | |
| GenBank mRNA | BC078790.1 | - | 1 - 2 | 115 | 7109515 | 143 |
| CB809217.1 | - | 1 - 2 | 149 | 143 | ||
| FQ230345.1 | - | 1 - 2 | 116 | 143 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC078790.1,NM_031670.2 transcripts.
Set Membership: |
|

Napsa
Genomic Map