Assay Details
| Gene Symbol: Nudt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007733.1 | NP_001007734.1 | 1 - 2 | 104 | 88 | |
| XM_008771875.1 | XP_008770097.1 | 1 - 2 | 123 | 88 | ||
| GenBank mRNA | BC079176.1 | - | 1 - 2 | 104 | 7130276 | 88 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079176.1, NM_001007733.1, XM_008771875.1 transcripts.
Set Membership: |
|

Nudt5
Genomic Map