Assay Details
| Gene Symbol: LOC685964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109489.1 | NP_001102959.1 | 2 - 3 | 201 | 131 | |
| XM_006236881.2 | XP_006236943.1 | 2 - 3 | 282 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC685964
Genomic Map