Assay Details
| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106004.2 | NP_001099474.1 | 1 - 2 | 77 | 65 | |
| GenBank mRNA | BC159414.1 | - | 1 - 2 | 77 | 8368074 | 65 |
| FQ212906.1 | - | 1 - 2 | 86 | 65 | ||
| FQ221260.1 | - | 1 - 2 | 78 | 65 | ||
More Information
Set Membership: |
|

Commd8
Genomic Map