Assay Details
| Gene Symbol: Dr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011914.1 | NP_001011914.1 | 1 - 2 | 843 | 101 | |
| GenBank mRNA | BC083822.1 | - | 1 - 2 | 843 | 7111461 | 101 |
| Gene Symbol: LOC100910207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751317.3 | XP_003751365.1 | 932 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dr1
Genomic Map