Assay Details
| Gene Symbol: Zc3h12d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107469.1 | NP_001100939.1 | 3 - 4 | 621 | 68 | |
| XM_006227616.2 | XP_006227678.1 | 3 - 4 | 1579 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zc3h12d
Genomic Map