Assay Details
| Gene Symbol: Zc3h12d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107469.1 | NP_001100939.1 | 5 - 6 | 964 | 68 | |
| XM_006227616.2 | XP_006227678.1 | 5 - 6 | 1922 | 68 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P081581 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P945723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12575 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P945723 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P081581 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P945723 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P081581 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P945723 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P081581 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P351537 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80552 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53012 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10701751 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701749 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701752 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701750 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10701748 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10701748 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21368976 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1649716 | - | - |

Zc3h12d
Genomic Map