Assay Details
| Gene Symbol: RGD1562084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109294.1 | NP_001102764.1 | 1169 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P092812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153520 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092812 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P092812 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P092812 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91254 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46570 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87737 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10910174 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910175 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910180 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910178 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910179 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910172 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910171 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910176 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910177 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10910173 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10910170 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800005075.rn.1 | TC0800000750.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10910170 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21711669 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368029 | - | - |

RGD1562084
Genomic Map