Assay Details
| Gene Symbol: RGD1562084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109294.1 | NP_001102764.1 | 181 | 72 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109294.1 transcripts.
Set Membership: |
|

RGD1562084
Genomic Map