Assay Details
| Gene Symbol: Pus7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729847.4 | XP_002729893.2 | 2 - 3 | 1013 | 79 | |
| XM_006242271.2 | XP_006242333.1 | 2 - 3 | 1320 | 79 | ||
More Information
Set Membership: |
|

Pus7l
Genomic Map