Assay Details
| Gene Symbol: Pqlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034952.1 | NP_001030124.1 | 1 - 2 | 314 | 79 | |
| XM_008764587.1 | XP_008762809.1 | 1 - 2 | 637 | 79 | ||
| GenBank mRNA | BC081879.1 | - | 1 - 2 | 220 | 7108131 | 79 |
| CB556892.1 | - | 1 - 2 | 468 | 79 | ||
| CK477351.1 | - | 1 - 2 | 137 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pqlc3
Genomic Map