Assay Details
| Gene Symbol: Pqlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034952.1 | NP_001030124.1 | 5 - 6 | 650 | 95 | |
| GenBank mRNA | CK477351.1 | - | 5 - 6 | 473 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pqlc3
Genomic Map