Assay Details
| Gene Symbol: Wdr35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099340.1 | NP_001092810.1 | 26 - 27 | 3470 | 86 | |
| XM_006239885.2 | XP_006239947.1 | 27 - 28 | 3519 | 86 | ||
| GenBank mRNA | EF613262.1 | - | 26 - 27 | 3470 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P029745 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P029740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144421 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029740 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029745 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029740 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P029745 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P029745 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P029740 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P380202 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70313 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81521 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10883580 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883563 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883561 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883572 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883567 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883558 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883570 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883566 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883569 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883564 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883574 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883579 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883582 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883562 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883573 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883575 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883560 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883576 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883565 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883578 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883571 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883559 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883577 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883581 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10883557 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600002801.rn.1 | TC0600000404.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002802.rn.1 | TC0600000404.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10883557 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22375837 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367773 | - | - |

Wdr35l
Genomic Map