Assay Details
| Gene Symbol: Adi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199097.1 | NP_954528.1 | 1 - 2 | 137 | 78 | |
| GenBank mRNA | AY346335.1 | - | 1 - 2 | 137 | 78 | |
| BC092562.1 | - | 1 - 2 | 126 | 7326245 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adi1
Genomic Map