Assay Details
| Gene Symbol: Isg20l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007741.1 | NP_001007742.1 | 1 - 2 | 240 | 101 | |
| GenBank mRNA | BC079314.1 | - | 1 - 2 | 240 | 7134903 | 101 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079314.1, NM_001007741.1 transcripts.
Set Membership: |
|

Isg20l2
Genomic Map