Assay Details
| Gene Symbol: Sptssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271299.1 | NP_001258228.1 | 2 - 3 | 633 | 73 | |
| GenBank mRNA | FM125939.1 | - | 2 - 3 | 150 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271299.1 transcripts.
Set Membership: |
|

Sptssb
Genomic Map