Assay Details
| Gene Symbol: Sptssb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271301.1 | NP_001258230.1 | 1 - 2 | 537 | 77 | |
| GenBank mRNA | BC087706.1 | - | 1 - 2 | 537 | 77 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271301.1 transcripts.
Set Membership: |
|

Sptssb
Genomic Map