Assay Details
| Gene Symbol: Cks1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135749.1 | NP_001129221.1 | 1 - 2 | 93 | 69 | |
| GenBank mRNA | BC167077.1 | - | 1 - 2 | 93 | 9023967 | 69 |
| BC168780.1 | - | 1 - 2 | 125 | 6918892 | 69 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cks1b
Genomic Map