Assay Details
| Gene Symbol: Dbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009644.1 | NP_001009644.1 | 3 - 4 | 667 | 83 | |
| GenBank mRNA | AB197138.1 | - | 3 - 4 | 667 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dbx1
Genomic Map