Assay Details
| Gene Symbol: LOC685463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001063889.4 | XP_001063889.2 | 485 | 93 | ||
| XM_008771169.1 | XP_008769391.1 | 611 | 93 | |||
| Gene Symbol: LOC100911649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751563.3 | XP_003751611.1 | 2 - 3 | 819 | 93 | |
| XM_008771171.1 | XP_008769393.1 | 1 - 2 | 945 | 93 | ||
| Gene Symbol: RGD1562641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_224663.7 | XP_224663.5 | 2 - 3 | 1743 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC685463
Genomic Map