Assay Details
| Gene Symbol: Grap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025749.1 | NP_001020920.1 | 3 - 4 | 370 | 68 | |
| GenBank mRNA | BC098740.1 | - | 3 - 4 | 370 | 7386199 | 68 |
| FQ228943.1 | - | 3 - 4 | 337 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grap
Genomic Map