Assay Details
| Gene Symbol: LOC102553035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229178.2 | XP_006229240.2 | 2 - 3 | 262 | 60 | |
| XM_008759447.1 | XP_008757669.1 | 3 - 4 | 242 | 60 | ||
| Gene Symbol: Klk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728759.4 | XP_002728805.2 | 3 - 4 | 407 | 60 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102553035
Genomic Map