Assay Details
| Gene Symbol: Mup5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203325.1 | NP_976070.1 | 279 | 81 | ||
| GenBank mRNA | AB039827.1 | - | 279 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P117653 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P214936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223853 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P214936 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117653 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P117653 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P214936 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P214936 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P117653 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P170502 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P214936 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10877231 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877253 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877259 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877375 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877283 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877351 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877258 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877238 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877315 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877239 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877296 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877311 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877361 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877349 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877373 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877261 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877322 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877364 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877284 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877219 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877243 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877316 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877347 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877319 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877321 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877228 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877320 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877374 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877345 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877227 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877323 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877310 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877242 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877355 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10877313 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10877217 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10877339 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10877278 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10877372 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10877307 | - | - |
| Rat Genome 230 2.0 Array | 1388031_x_at | - | - |
| Rat Genome 230 2.0 Array | 1370779_PM_x_at | - | - |
| Rat Genome 230 2.0 Array | 1370778_at | - | - |
| Rat Genome 230 2.0 Array | 1370778_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370779_x_at | - | - |
| Rat Genome 230 2.0 Array | 1388031_PM_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1370778_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370779_PM_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1370778_at | - | - |
| Affymetrix HT Rat Focus Array | 1370779_x_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500021355.rn.1 | TC0500002863.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500021370.rn.1 | TC0500002863.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500021366.rn.1 | TC0500002863.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500021371.rn.1 | TC0500002863.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500021372.rn.1 | TC0500002863.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388031_PM_x_at | - | - |
| Rat Expression Set 230 | 1388031_x_at | - | - |
| Rat Expression Set 230 | 1370779_x_at | - | - |
| Rat Expression Set 230 | 1370779_PM_x_at | - | - |
| Rat Expression Set 230 | 1370778_PM_at | - | - |
| Rat Expression Set 230 | 1370778_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877372 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877251 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877339 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877286 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877229 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877313 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877307 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10877295 | - | - |
| Rat Genome U34 Set | M24108mRNA_at | - | - |
| Rat Genome U34 Set | M24109mRNA_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21580420 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355639 | - | - |
| RatRef-12 Expression BeadChip | ILMN_48339 | - | - |

Mup5
Genomic Map