Assay Details
| Gene Symbol: Ces2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271303.1 | NP_001258232.1 | 10 - 11 | 1470 | 167 | |
| Gene Symbol: LOC100365112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190369.1 | NP_001177298.1 | 10 - 11 | 1472 | 167 | |
| GenBank mRNA | AB010632.1 | - | 10 - 11 | 1442 | 167 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Ces2
Genomic Map