Assay Details
| Gene Symbol: Cd24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012752.3 | NP_036884.1 | 2 | 953 | 135 | |
| GenBank mRNA | BC064439.1 | - | 2 | 937 | 5623514 | 135 |
| BC081851.1 | - | 2 | 948 | 7110912 | 135 | |
| Z11663.1 | - | 2 | 892 | 135 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cd24
Genomic Map