Assay Details
| Gene Symbol: Gjb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053388.1 | NP_445840.1 | 3 | 1616 | 142 | |
| XM_006252079.2 | XP_006252141.1 | 3 | 2942 | 142 | ||
| XM_006252080.2 | XP_006252142.1 | 3 | 2518 | 142 | ||
| XM_008770781.1 | XP_008769003.1 | 3 | 1730 | 142 | ||
| XM_008770782.1 | XP_008769004.1 | 3 | 2040 | 142 | ||
| XM_008770783.1 | XP_008769005.1 | 3 | 1621 | 142 | ||
| XM_008770784.1 | XP_008769006.1 | 3 | 1727 | 142 | ||
| GenBank mRNA | AF170284.1 | - | 3 | 1616 | 142 | |
| BC078753.1 | - | 3 | 1613 | 7132296 | 142 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gjb6
Genomic Map