Assay Details
| Gene Symbol: Elfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105913.1 | NP_001099383.1 | 2 | 4040 | 122 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P163574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46113 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P163574 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P163574 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P163574 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P102698 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10760645 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10760644 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200010074.rn.1 | TC1200001186.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200010079.rn.1 | TC1200001186.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200010075.rn.1 | TC1200001186.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200010080.rn.1 | TC1200001186.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10760644 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21297568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650129 | - | - |

Elfn1
Genomic Map