Assay Details
| Gene Symbol: Hoxd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271038.1 | NP_001257967.1 | 3 | 2443 | 131 | |
| XM_008761880.1 | XP_008760102.1 | 2 | 2703 | 131 | ||
| XM_008761881.1 | XP_008760103.1 | 3 | 2181 | 131 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hoxd3
Genomic Map