Assay Details
| Gene Symbol: Traf3ip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044248.1 | NP_001037713.1 | 4 | 1214 | 149 | |
| XM_006256532.2 | XP_006256594.1 | 4 | 1431 | 149 | ||
| XM_006256533.2 | XP_006256595.1 | 3 | 926 | 149 | ||
| GenBank mRNA | BC100099.1 | - | 4 | 1214 | 7376204 | 149 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26008 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14795 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14795 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14795 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14795 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21247 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13377 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34911 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59359 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22463 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10830400 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830403 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830407 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830406 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830401 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830405 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830399 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830404 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830402 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10830398 | - | - |
| Rat Genome 230 2.0 Array | 1374671_at | - | - |
| Rat Genome 230 2.0 Array | 1374671_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374671_at | - | - |
| Affymetrix HT Rat Focus Array | 1374671_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000006696.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006690.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006698.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006693.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006697.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006682.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006694.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006704.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006691.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006703.rn.1 | TC2000000797.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR2000006702.rn.1 | TC2000000797.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374671_at | - | - |
| Rat Expression Set 230 | 1374671_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10830398 | - | - |
| Rat Genome U34 Set | rc_AA892179_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21843783 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366602 | - | - |

Traf3ip2
Genomic Map