Assay Details
| Gene Symbol: MGC109340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024267.2 | NP_001019438.1 | 1 | 1453 | 78 | |
| GenBank mRNA | BC092634.1 | - | 1 | 1455 | 7315105 | 78 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MGC109340
Genomic Map