Assay Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250316.2 | XP_006250378.1 | 5 | 4319 | 123 | |
| XM_006250317.2 | XP_006250379.1 | 6 | 4733 | 123 | ||
| XM_008769783.1 | XP_008768005.1 | 5 | 4276 | 123 | ||
| XM_008769784.1 | XP_008768006.1 | 6 | 4452 | 123 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Desi2
Genomic Map