Assay Details
| Gene Symbol: H2afy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135807.1 | NP_001129279.1 | 4 | 602 | 103 | |
| GenBank mRNA | BC161981.1 | - | 4 | 602 | 9029999 | 103 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

H2afy2
Genomic Map