Assay Details
| Gene Symbol: Mafg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247912.2 | XP_006247974.1 | 1 | 112 | ||
| XM_006247916.2 | XP_006247978.1 | 1 | 112 | |||
| XM_006247917.2 | XP_006247979.1 | 1 | 112 | |||
| XM_006247918.2 | XP_006247980.1 | 1 | 112 | |||
| XM_006247919.2 | XP_006247981.1 | 1 | 112 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_006247912.2,XM_006247916.2,XM_006247917.2,XM_006247918.2,XM_006247919.2 transcripts.
This assay is in the 5'UTR region of the XM_006247912.2,XM_006247916.2,XM_006247917.2,XM_006247918.2,XM_006247919.2 transcripts.
Set Membership: |
|

Mafg
Genomic Map