Assay Details
| Gene Symbol: Mpeg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231115.2 | XP_006231177.1 | 2 | 3924 | 140 | |
| XM_008760261.1 | XP_008758483.1 | 2 | 4016 | 140 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mpeg1
Genomic Map