Assay Details
| Gene Symbol: Olr1700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001113.1 | NP_001001113.1 | 1 | 780 | 86 | |
| Gene Symbol: LOC103689944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255858.2 | XP_006255920.1 | 1 | 618 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Olr1700
Genomic Map