Assay Details
| Gene Symbol: RGD1311805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009638.1 | NP_001009638.1 | 17 - 18 | 1679 | 153 | |
| GenBank mRNA | BC087672.1 | - | 17 - 18 | 1679 | 7317049 | 153 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311805
Genomic Map