Assay Details
| Gene Symbol: Ackr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078621.2 | NP_511176.2 | 1 | 1634 | 124 | |
| XM_008766664.1 | XP_008764886.1 | 3 | 2152 | 124 | ||
| XM_008766665.1 | XP_008764887.1 | 3 | 2493 | 124 | ||
| GenBank mRNA | BC086449.1 | - | 1 | 1634 | 7304127 | 124 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ackr2
Genomic Map