Assay Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013873.1 | NP_001013895.1 | 3 - 4 | 609 | 101 | |
| XM_006250316.2 | XP_006250378.1 | 3 - 4 | 959 | 101 | ||
| XM_006250317.2 | XP_006250379.1 | 4 - 5 | 1373 | 101 | ||
| XM_008769783.1 | XP_008768005.1 | 3 - 4 | 916 | 101 | ||
| XM_008769784.1 | XP_008768006.1 | 4 - 5 | 1092 | 101 | ||
| XM_008769785.1 | XP_008768007.1 | 3 - 4 | 1220 | 101 | ||
| GenBank mRNA | BC083584.1 | - | 3 - 4 | 609 | 7131107 | 101 |
More Information
Updated Mapping Information
Set Membership: |
|

Desi2
Genomic Map