Assay Details
| Gene Symbol: Dyrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024767.1 | NP_001019938.1 | 2 - 3 | 299 | 60 | |
| XM_008769464.1 | XP_008767686.1 | 2 - 3 | 511 | 60 | ||
| GenBank mRNA | BC097474.1 | - | 2 - 3 | 299 | 7457808 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Dyrk3
Genomic Map