Assay Details
| Gene Symbol: Apon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009385.1 | NP_001009385.1 | 2 | 1053 | 109 | |
| GenBank mRNA | BC088168.1 | - | 2 | 1053 | 7322067 | 109 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Apon
Genomic Map